4-tert-Amylphenol ≥99%

Synonyms: p-(1,1-Dimethylpropyl)phenol, 4-(2-Methyl-2-butyl)phenol, 4-tert-Pentylphenol

153842-5G 153842-100G
220037-371EA 48.25 USD
220037-371 220037-370
4-tert-Amylphenol ≥99%
4-tert-Amylphenol
Formula: C₁₁H₁₆O
MW: 164.25 g/mol
MDL Number: MFCD00002369
CAS Number: 80-46-6
EINECS: 201-280-9
UN: 2430
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR