3,5-Dichlorophenylhydrazinium chloride ≥95%

Synonyms: 1-(3,5-Dichlorophenyl)hydrazine hydrochloride, 3,5-Dichlorophenylhydrazine hydrochloride

153648-10G 153648-50G
220038-020EA 79.26 USD
220038-020 220038-021
3,5-Dichlorophenylhydrazinium chloride ≥95%
3,5-Dichlorophenylhydrazinium chloride
Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
MDL Number: MFCD00012938
CAS Number: 63352-99-8
EINECS: 264-109-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR