1-(Trimethylsilyl)imidazole

153583-100ML 153583-25ML 153583-5ML
220038-010EA 284.31 USD
220038-010 220038-011 220038-012
1-(Trimethylsilyl)imidazole
1-(Trimethylsilyl)imidazole
Formula: C₆H₁₂N₂Si
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: –42 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: 61 °C
MDL Number: MFCD00005280
CAS Number: 18156-74-6
EINECS: 242-040-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR