2-Fluorophenylhydrazinium chloride ≥97%

Synonyms: (2-Fluorophenyl)hydrazine hydrochloride, (2-Fluorphenyl)hydrazin HCl, 1-(2-Fluorophenyl)hydrazine hydrochloride, 2-Fluorophenylhydrazine hydrochloride

153427-25G 153427-5G
220037-991EA 85.72 USD
220037-991 220037-992
2-Fluorophenylhydrazinium chloride ≥97%
2-Fluorophenylhydrazinium chloride
Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
MDL Number: MFCD00012927
CAS Number: 2924-15-4
EINECS: 220-885-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR