2,6-Dichlorophenylhydrazinium chloride ≥98%

Synonyms: 2,6-Dichlorophenylhydrazine hydrochloride, 1-(2,6-Dichlorophenyl)hydrazine hydrochloride

153419-1G 153419-10G
220037-989EA 31.26 USD
220037-989 220037-988
2,6-Dichlorophenylhydrazinium chloride ≥98%
2,6-Dichlorophenylhydrazinium chloride
Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
MDL Number: MFCD00012930
CAS Number: 50709-36-9
EINECS: 256-730-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR