1-Bromo-4-chloro-2-nitrobenzene ≥97%

Warning

153184-100G 153184-25G
220037-969EA 34.49 USD
220037-969 220037-970
1-Bromo-4-chloro-2-nitrobenzene ≥97%
1-Bromo-4-chloro-2-nitrobenzene
Formula: C₆H₃BrClNO₂
MW: 236.45 g/mol
Melting Pt: 70 °C
Density: 2.048 g/cm³ (20 °C)
MDL Number: MFCD00024320
CAS Number: 41513-04-6
EINECS: 255-421-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR