3,4-Dichlorophenylhydrazinium chloride ≥90%, Technical Grade

Synonyms: 3,4-Dichlorophenylhydrazine hydrochloride, 1-(3,4-Dichlorophenyl)hydrazine hydrochloride

152811-25G 152811-100G
220037-890EA 248.77 USD
220037-890 220037-889
3,4-Dichlorophenylhydrazinium chloride ≥90%, Technical Grade
3,4-Dichlorophenylhydrazinium chloride
Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
MDL Number: MFCD00012937
CAS Number: 19763-90-7
EINECS: 243-278-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR