1,4-Naphthoquinone

Danger

152757-5G 152757-100G 152757-500G
220037-885EA 53.42 USD
220037-885 220037-243 220037-244
1,4-Naphthoquinone
1,4-Naphthoquinone
Formula: C₁₀H₆O₂
MW: 158.16 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 124 °C
Density: 1.422 g/cm³ (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00001676
CAS Number: 130-15-4
EINECS: 204-977-6
UN: 2811
ADR: 6.1,I
Merck Index: 12,06482

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR