2-Chloro-6-methoxypyridine

Warning

152730-5G 152730-25G
220037-242EA 29.08 USD
220037-242 220037-241
2-Chloro-6-methoxypyridine
2-Chloro-6-methoxypyridine
Formula: C₆H₆ClNO
MW: 143.57 g/mol
Boiling Pt: 39…40 °C (0.3 mmHg)
Density: 1.207 g/cm³ (20 °C)
MDL Number: MFCD00006265
CAS Number: 17228-64-7
EINECS: 241-264-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR