4-Bromo-α,α,α-trifluorotoluene ≥99%

Danger

Synonyms: 4-Bromobenzotrifluoride, 1-Bromo-4-(trifluoromethyl)benzene

152692-5G 152692-100G 152692-25G
220037-236EA 46.31 USD
220037-236 220037-234 220037-235
4-Bromo-α,α,α-trifluorotoluene ≥99%
4-Bromo-α,α,α-trifluorotoluene
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 154 °C (1013 hPa)
Density: 1.6347 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000398
CAS Number: 402-43-7
EINECS: 206-943-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR