Sodium toluene-4-sulfonate ≥95%

Synonyms: p-Toluenesulphonic acid sodium salt, Sodium p-toluenesulphonate, Toluene-4-sulphonic acid sodium salt

152536-500G 152536-5G 152536-100G
220037-883EA 155.08 USD
220037-883 220037-884 220037-882
Sodium toluene-4-sulfonate ≥95%
Sodium toluene-4-sulphonate
Formula: C₇H₇NaO₃S
MW: 194.19 g/mol
MDL Number: MFCD00798566
CAS Number: 657-84-1
EINECS: 211-522-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR