4-Methoxy-3-methylbenzaldehyde ≥99%

Synonyms: 3-Methyl-4-anisaldehyde

152129-50G 152129-10G
220037-847EA 1346.15 USD
220037-847 220037-846
4-Methoxy-3-methylbenzaldehyde ≥99%
4-Methoxy-3-methylbenzaldehyde
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 80 °C (1013 hPa)
Density: 1.082 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003375
CAS Number: 32723-67-4
EINECS: 251-177-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR