1-Isoquinolinol ≥98%

152102-1G 152102-5G 152102-10G
220037-843EA 68.17 USD
220037-843 220037-844 220037-842
1-Isoquinolinol ≥98%
1-Isoquinolinol
Formula: C₉H₇NO
Storage Temperature: Ambient
MDL Number: MFCD00006899
CAS Number: 491-30-5
EINECS: 207-732-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR