Trifluoroacetic acid-[D₁]

Danger

152005-25G 152005-10G 152005-5G
220037-829EA 46.46 USD
220037-829 220037-612 220037-830
Trifluoroacetic acid-[D₁]
Trifluoroacetic acid-[D₁]
Formula: F₃CCOOD
Boiling Pt: 75 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00044563
CAS Number: 599-00-8
EINECS: 209-961-2
UN: 2699
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR