n-Octane-[D₁₈]

Danger

Synonyms: n-Octane-[²H₁₈], Octane-D18

151971-1G 151971-5G
220037-605EA 45.85 USD
220037-605 220037-606
n-Octane-[D₁₈]
n-Octane-[D₁₈]
Formula: D₃C(CD₂)₆CD₃
MW: 132.09 g/mol
Boiling Pt: 125…126 °C (1013 hPa)
Melting Pt: –57 °C
Density: 0.81 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00037626
CAS Number: 17252-77-6
EINECS: 241-285-3
UN: 1262
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR