Nitrobenzene-[D₅]

Synonyms: Nitrobenzene-[²H₅]

151955-25G 151955-5G 151955-10G
220037-600EA 68.62 USD
220037-600 220037-601 220037-599
Nitrobenzene-[D₅]
Nitrobenzene-[D₅]
Formula: C₆D₅NO₂
MW: 128.07 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: 5 °C
Density: 1.25 g/cm³ (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00044415
CAS Number: 4165-60-0
EINECS: 224-014-3
UN: 1662
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR