Benzene-[D₆]

Danger

151815-100G 151815-25G 151815-50G
220037-788EA 152.31 USD
220037-788 220037-790 220037-791
Benzene-[D₆]
Benzene-[D₆]
Formula: C₆D₆
Boiling Pt: 79 °C (1013 hPa)
Melting Pt: 6.7 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: –11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003010
CAS Number: 1076-43-3
EINECS: 214-061-8
UN: 1114
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR