Isoxazole

151637-25G 151637-5G
220037-757EA 117.38 USD
220037-757 220037-758
Isoxazole
Isoxazole
Formula: C₃H₃NO
MW: 69.06 g/mol
Boiling Pt: 95 °C (1013 hPa)
Density: 1.08 g/cm³ (20 °C)
Flash Pt: 10 °C
MDL Number: MFCD00003149
CAS Number: 288-14-2
EINECS: 206-018-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR