6-Chloro-m-anisidine hydrochloride ≥96%

Synonyms: 2-Chloro-5-methoxyanilinium chloride, 3-Amino-4-chloroanisole hydrochloride, 2-Chloro-5-methoxyaniline hydrochloride, 6-Chloro-3-anisidine hydrochloride

151602-5G 151602-25G
220037-752EA 666.09 USD
220037-752 220037-751
6-Chloro-m-anisidine hydrochloride ≥96%
6-Chloro-m-anisidine hydrochloride
Formula: C₇H₉Cl₂NO
MW: 194.06 g/mol
Melting Pt: 208 °C
MDL Number: MFCD00012962
CAS Number: 85006-21-9
EINECS: 285-050-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR