Trichloroacetyl chloride ≥99%

Danger

151599-500G 151599-100G 151599-25G
220037-749EA 302.62 USD
220037-749 220037-747 220037-748
Trichloroacetyl chloride ≥99%
Trichloroacetyl chloride
Formula: C₂Cl₄O
MW: 181.83 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: -57 °C
Density: 1.62 g/cm³ (20 °C)
MDL Number: MFCD00000792
CAS Number: 76-02-8
EINECS: 200-926-7
UN: 2442
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR