3-Nitro-p-toluic acid ≥99%

Synonyms: 4-Methyl-3-nitrobenzoic acid

151408-100G 151408-25G
220037-723EA 66.23 USD
220037-723 220037-724
3-Nitro-p-toluic acid ≥99%
3-Nitro-p-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 360 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00007174
CAS Number: 96-98-0
EINECS: 202-549-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR