6-Nitro-o-toluic acid ≥97%

Synonyms: 2-Methyl-6-nitrobenzoic acid

151394-25G 151394-5G
220037-718EA 280.25 USD
220037-718 220037-719
6-Nitro-o-toluic acid ≥97%
6-Nitro-o-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 353 °C (1013 hPa)
Melting Pt: 151…153 °C
MDL Number: MFCD00007267
CAS Number: 13506-76-8
EINECS: 236-833-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR