Fumaroyl chloride ≥95%

Synonyms: Fumaryl chloride

151386-5G 151386-25G 151386-100G
220037-716EA 39 USD
220037-716 220037-715 220037-714
Fumaroyl chloride ≥95%
Fumaroyl chloride
Formula: C₄H₂Cl₂O₂
MW: 152.96 g/mol
Boiling Pt: 161 °C (1013 hPa)
Melting Pt: –2 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000733
CAS Number: 627-63-4
EINECS: 211-005-4
UN: 1780
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR