Bis(4-chlorophenyl) sulfone ≥98%

Synonyms: 4,4'-Dichloro diphenyl sulphone, 4-Chlorophenyl sulphone, 4,4'-sulphonylbis(chlorobenzene)

151378-500G 151378-100G
220037-712EA 332.77 USD
220037-712 220037-711
Bis(4-chlorophenyl) sulfone ≥98%
Bis(4-chlorophenyl)sulphone
Formula: C₁₂H₈Cl₂O₂S
MW: 287.17 g/mol
MDL Number: MFCD00000619
CAS Number: 80-07-9
EINECS: 201-247-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR