1-(2-Pyridyl)piperazine

Synonyms: 2-(1-Piperazinyl)pyridine

151270-25G 151270-5G
220037-704EA 46.31 USD
220037-704 220037-705
1-(2-Pyridyl)piperazine
1-(2-Pyridyl)piperazine
Formula: C₉H₁₃N₃
MW: 163.22 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 85…87 °C
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00006216
CAS Number: 34803-66-2
EINECS: 252-220-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR