2,5-Dimethylbenzaldehyde ≥99%

151068-25G 151068-5G
220037-698EA 586.92 USD
220037-698 220037-699
2,5-Dimethylbenzaldehyde ≥99%
2,5-Dimethylbenzaldehyde
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 104.4 °C (1013 hPa)
Density: 0.95 g/cm³ (20 °C)
MDL Number: MFCD00003342
CAS Number: 5779-94-2
EINECS: 227-303-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR