6-Bromocaproic acid ≥97%

Danger

150452-25G 150452-100G 150452-5G
220037-646EA 76.57 USD
220037-646 220037-645 220037-647
6-Bromocaproic acid ≥97%
6-Bromocaproic acid
Formula: BrH₂C(CH₂)₄COOH
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: 31…34 °C
Density: 1.4068 g/cm³ (30 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004422
CAS Number: 4224-70-8
EINECS: 224-176-5
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR