4-tert-Butylbenzoic acid ≥99%

Synonyms: 4-(tert-Butyl)benzoic acid

150355-500G 150355-2KG
220036-921EA 93.69 USD
220036-921 220036-920
4-tert-Butylbenzoic acid ≥99%
4-tert-Butylbenzoic acid
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 165 - 167 °C
Density: 0.6 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00002563
CAS Number: 98-73-7
EINECS: 202-696-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR