4-Chloroquinaldine ≥99%

Warning

150231-25G 150231-5G
220037-321EA 384.23 USD
220037-321 220037-322
4-Chloroquinaldine ≥99%
4-Chloroquinaldine
Formula: C₁₀H₈ClN
MW: 177.63 g/mol
MDL Number: MFCD00006757
CAS Number: 4295-06-1
EINECS: 224-300-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR