1-Isoquinolinecarboxylic acid ≥99%

Synonyms: Isoquinoline-1-carboxylic acid

150134-5G 150134-25G
220037-579EA 98.86 USD
220037-579 220037-578
1-Isoquinolinecarboxylic acid ≥99%
1-Isoquinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD00006901
CAS Number: 486-73-7
EINECS: 207-639-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR