2,4-Dimethyl-3-pentanol ≥99%

Synonyms: 2,4-Dimethylpentan-3-ol

150045-25G 150045-100G
220037-569EA 41.68 USD
220037-569 220037-568
2,4-Dimethyl-3-pentanol ≥99%
2,4-Dimethyl-3-pentanol
Formula: C₇H₁₆O
MW: 116.2 g/mol
Boiling Pt: 139 °C (1013 hPa)
Melting Pt: -70 °C
Density: 0.8445 g/cm³ (0 °C)
MDL Number: MFCD00008921
CAS Number: 600-36-2
EINECS: 209-993-7
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR