3-Chloro-o-anisidine ≥97%

Synonyms: 3-Chloro-2-methoxyaniline

149942-5G 149942-1G
220035-756EA 233.69 USD
220035-756 220035-755
3-Chloro-o-anisidine ≥97%
3-Chloro-o-anisidine
Formula: C₇H₈ClNO
MW: 157.6 g/mol
MDL Number: MFCD00060607
CAS Number: 51114-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR