Thallium(I) ethanolate

Synonyms: Thallium (I) ethoxide

149845-5G 149845-25G
220037-550EA 28.32 USD
220037-550 220036-554
Thallium(I) ethanolate
Thallium(I) ethanolate
Formula: C₂H₅OTl
MW: 249.44 g/mol
Melting Pt: -3 °C
Density: 3.52 g/cm³ (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00009072
CAS Number: 20398-06-5
EINECS: 243-786-2
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR