Di-isopropanolamine ≥98.0% (by titrimetric analysis)

Warning

Synonyms: 1,1'-Iminodipropan-2-ol, Bis(2-hydroxypropyl)amine, 1,1-Iminodi-2-propanol, 1-[(2-Hydroxypropyl)amino]propan-2-ol

14960-1KG 14960-250G
220037-544EA 135.69 USD
220037-544 220037-545
Di-isopropanolamine ≥98.0% (by titrimetric analysis)
Di-isopropanolamine
Formula: C₆H₁₅NO₂
MW: 133.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004531
CAS Number: 110-97-4
EINECS: 203-820-9

Order Now

Specification Test Results

Assay 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR