2,5-Dihydroxybenzoic acid ≥98%

Synonyms: 2,5-Dihydroxybenzoic Acid, DHB, Hydroquinonecarboxylic acid, Gentisic acid, 2,5-DHBA

149357-100G 149357-25G 149357-10G
220037-531EA 192.77 USD
220037-531 220037-533 220037-532
2,5-Dihydroxybenzoic acid ≥98%
2,5-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 202…206 °C
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002460
CAS Number: 490-79-9
EINECS: 207-718-5
Merck Index: 13,04409

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR