4-(Dimethylamino)benzophenone ≥98%

149349-10G 149349-25G
220037-529EA 244.54 USD
220037-529 220037-530
4-(Dimethylamino)benzophenone ≥98%
4-(Dimethylamino)benzophenone
Formula: C₁₅H₁₅NO
MW: 225.29 g/mol
Boiling Pt: 245…250 °C (15 mmHg)
Melting Pt: 97…98 °C
MDL Number: MFCD00008311
CAS Number: 530-44-9
EINECS: 208-478-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR