Ethyl decanoate ≥99%

Synonyms: Ethyl caprate, Capric acid ethyl ester, Decanoic acid ethyl ester, Decylic acid ethyl ester

148970-500ML 148970-100ML
220037-514EA 119.54 USD
220037-514 220037-513
Ethyl decanoate ≥99%
Ethyl decanoate
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: -20 °C
Density: 0.862 g/cm³ (20 °C)
MDL Number: MFCD00009581
CAS Number: 110-38-3
EINECS: 203-761-9
Merck Index: 13,03811

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR