2-Amino-3-hydroxybenzoic acid ≥97%

Synonyms: 3-Hydroxyanthranilic acid

148776-1G 148776-250MG
220036-963EA 1303.08 USD
220036-963 220036-964
2-Amino-3-hydroxybenzoic acid ≥97%
2-Amino-3-hydroxybenzoic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 164 °C
MDL Number: MFCD00007700
CAS Number: 548-93-6
EINECS: 208-962-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR