Thiobenzamide ≥98%

Synonyms: Benzothioamide

148229-10G 148229-50G
220037-493EA 23.31 USD
220037-493 220037-494
Thiobenzamide ≥98%
Thiobenzamide
Formula: C₇H₇NS
MW: 137.21 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 113…116 °C
MDL Number: MFCD00008060
CAS Number: 2227-79-4
EINECS: 218-765-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR