4-Methylanisole ≥99%

Warning

Synonyms: p-Methylanisole

148091-100ML 148091-1L
220037-485EA 26.38 USD
220037-485 220037-486
4-Methylanisole ≥99%
4-Methylanisole
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: –36 °C
Density: 0.97 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00008413
CAS Number: 104-93-8
EINECS: 203-253-7
UN: 3271
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR