1,1,1-Tris(hydroxymethyl)propane ≥97%

Synonyms: 1,1,1-Trimethylolpropane, Propylidynetrimethanol, Trimethylol propane, Propylidynetris(methanol), 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol

148083-3KG 148083-500G
220037-483EA 135.69 USD
220037-483 220037-484
1,1,1-Tris(hydroxymethyl)propane ≥97%
1,1,1-Tris(hydroxymethyl)propane
Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 53 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004694
CAS Number: 77-99-6
EINECS: 201-074-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR