DL-Camphor

Warning

Synonyms: (±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (±)-1,7,7-Trimethyl-2-norbornanone

148075-500G 148075-100G
220037-482EA 81.09 USD
220037-482 220037-481
DL-Camphor
DL-Camphor
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: ~ 180 °C
Density: 0.992 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR