Methyl isobutyrate ≥99%

148008-500ML 148008-100ML
220037-477EA 99.4 USD
220037-477 220037-476
Methyl isobutyrate ≥99%
Methyl isobutyrate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
MDL Number: MFCD00008914
CAS Number: 547-63-7
EINECS: 208-929-5
UN: 1237
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR