(R)-(-)-2-Octanol ≥99%

Warning

Synonyms: (R)-(-)-2-Hydroxy octane, (R)-2-Octanol, L(-)-2-Octanol, (R)-(-)-Hexyl methyl carbinol, (R)-(-)-Methyl hexyl carbinol

147990-10G 147990-5G
220037-474EA 402.23 USD
220037-474 220037-475
(R)-(-)-2-Octanol ≥99%
(R)-(-)-2-Octanol
Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 175 °C (1013 hPa)
Density: 0.8201 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064284
CAS Number: 5978-70-1
EINECS: 227-777-0
UN: 1987
ADR: 3,III
Merck Index: 12,06850
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18fd46b3d9d8ab0ee6af423314fb7b52b40bdfc5&vid=84b3ca36a2d8e2a6ae990fc18ec360718d483dc0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR