DL-α-Phenylalanine ≥99%

Synonyms: H-DL-Phe-OH, DL-Phenylalanine, 3-Phenyl-DL-alanine

147966-100G 147966-25G
220037-470EA 75.6 USD
220037-470 220037-471
DL-α-Phenylalanine ≥99%
DL-α-Phenylalanine
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 173 °C
MDL Number: MFCD00064225
CAS Number: 150-30-1
EINECS: 205-756-7
Merck Index: 13,07355

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR