4,4'-Bis(dimethylamino)benzophenone

Danger

Synonyms: 4,4′-Bis(dimethylamino)benzophenone, 4,4-Bis(dimethylamino)benzophenone, 4,4'-Di(dimethylamino)benzophenone, Michler's ketone, Bis[4-(dimethylamino)phenyl]methanone

147834-100G 147834-500G
220037-456EA 68 USD
220037-456 220037-457
4,4'-Bis(dimethylamino)benzophenone
4,4'-Bis(dimethylamino)benzophenone
Formula: C₁₇H₂₀N₂O
MW: 268.36 g/mol
Boiling Pt: 428 °C (1013 hPa)
Melting Pt: 172 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00008312
CAS Number: 90-94-8
EINECS: 202-027-5
UN: 2811
ADR: 6.1,III
Merck Index: 12,06265

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR