5,6-Dimethoxy-1-indanone

Synonyms: 5,6-Dimethoxy-2,3-dihydro-1H-inden-1-one

147826-25G 147826-5G
220037-454EA 97.25 USD
220037-454 220037-455
5,6-Dimethoxy-1-indanone
5,6-Dimethoxy-1-indanone
Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00003790
CAS Number: 2107-69-9
EINECS: 218-287-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR