Diethyl cyclopropane-1,1-dicarboxylate ≥97%

Synonyms: Diethyl 1,1-cyclopropanedicarboxylate, Diethyl 1,1-cyclopropane dicarboxyliate

147710-25G 147710-5G
220037-447EA 316.15 USD
220037-447 220037-448
Diethyl cyclopropane-1,1-dicarboxylate ≥97%
Diethyl cyclopropane-1,1-dicarboxylate
Formula: C₉H₁₄O₄
MW: 186.21 g/mol
MDL Number: MFCD00001283
CAS Number: 1559-02-0
EINECS: 216-321-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR