Benzene-1,3,5-tricarbonyl trichloride ≥98%

Danger

Synonyms: 1,3,5-Benzenetricarboxylic acid chloride, 1,3,5-Benzenetricarbonyl chloride, Trimesoylchloride, Trimesoyl chloride

147532-12X500G 147532-2.5KG 147532-25G 147532-100G 147532-10G 147532-500G
220037-117EA 69.37 USD
220037-117 220037-118 220037-119 220037-115 220037-116 220037-120
Benzene-1,3,5-tricarbonyl trichloride ≥98%
Benzene-1,3,5-tricarbonyl trichloride
Formula: C₉H₃Cl₃O₃
MW: 265.48 g/mol
Boiling Pt: 312 °C (1013 hPa)
Melting Pt: 34…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00000679
CAS Number: 4422-95-1
EINECS: 224-594-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR