trans-1,2-Cyclohexanedicarboxylic acid ≥95%

Synonyms: trans-Cyclohexane-1,2-dicarboxylic acid

147516-5G 147516-25G
220037-111EA 509.05 USD
220037-111 220037-110
trans-1,2-Cyclohexanedicarboxylic acid ≥95%
trans-1,2-Cyclohexanedicarboxylic acid
Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 220…221 °C
MDL Number: MFCD00001464
CAS Number: 2305-32-0
EINECS: 218-975-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR